Key Documents
TEM-CFLF400CU
Sigma-Aldrich
Carbon Film Supported Copper London Finder LF400 Grids
box of 25
Synonym(s):
CARBON ONLY ON LONDON FINDER 400# COPPER GRIDS 25/BX
About This Item
UNSPSC Code:
41111761
packaging
box of 25
General description
A 5-6 nm thin film of pure carbon supported London finder (LF400) grids. Copper grid. Carbon film show better electrical and heat conduction in the electron beam. London finder grids are locator grids with reference/index on the grid.
Choose from one of the most recent versions:
Certificates of Analysis (COA)