Key Documents
TEM-CFGA75CU
Sigma-Aldrich
Carbon Film Supported Copper Single Hole Grid
hole size 75 μm, box of 25
Synonym(s):
CARBON FILM ONLY SINGLE HOLE 75 MICRON COPPER GRIDS 25/BX
About This Item
UNSPSC Code:
41111761
packaging
box of 25
hole size
75 μm
General description
A 5-6 nm thin film of pure carbon supported single hole grid. Copper grid, 75 micron hole. Carbon film show better electrical and heat conduction in the electron beam.
Choose from one of the most recent versions:
Certificates of Analysis (COA)