Key Documents
TEM-CF400CU50
Sigma-Aldrich
Carbon Film Supported Copper Grid
Standard Thickness, size 400 mesh, box of 50
Synonym(s):
Carbon Film 400 Mesh, Copper
About This Item
UNSPSC Code:
41111761
material
carbon film
copper grid
packaging
box of 50
film size
5-6 nm
size
400 mesh
General description
TEM grids supported by carbon film only. A 5-6 nm thin film of pure carbon deposited on one side of the standard square mesh grid.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Questions
1–2 of 2 Questions  
  1. Where is the difference between product 931500 and this product TEM-CF400CU50?
    1 answer
    1. There are no major differences between product TEM-CF400CU50 and 931500. Product 931500 is a direct alternative to product TEM-CF400CU50.
      Helpful?
  2. Hello, I need to purchase carbon TEM grids. Are these the same as the ones from Ted Pella? Product Number: 01844-F Carbon Film only on 400 mesh, Copper
    1 answer
    1. It appears that the description and specifications of Product TEM-CF400CU50 are similar to the Ted Pella carbon TEM grids, and may be considered as a suitable alternative to this product. It is the end user's responsibility to determine the suitability of the product for their application.
      Helpful?