Key Documents
TEM-CF300HCU
Sigma-Aldrich
Carbon Film Supported Copper Hexagonal Mesh Grid
size 300 mesh, box of 25
Synonym(s):
CARBON FILM ONLY ON 300 HEX COPPER GRIDS 25/BX
About This Item
UNSPSC Code:
41111761
material
carbon film
copper grid
description
film layer thickness 5 - 6 nm carbon
packaging
box of 25
size
300 mesh
General description
A 5-6 nm thin film of pure carbon supported hexagonal mesh grids. Copper grid, 300 mesh. Carbon film show better electrical and heat conduction in the electron beam.
Choose from one of the most recent versions:
Certificates of Analysis (COA)