Key Documents
TEM-CF300AU50
Sigma-Aldrich
Carbon Film Supported Gold Square Mesh
size 300 mesh, box of 50
Synonym(s):
CARBON FILM 300 MESH AU 50/BX
About This Item
UNSPSC Code:
41111761
packaging
box of 50
size
300 mesh
General description
A 5-6 nm thin film of pure carbon supported standard squre mesh grids. Gold grid, 300 mesh.
Choose from one of the most recent versions:
Certificates of Analysis (COA)