Key Documents
TEM-CF200HCU50
Sigma-Aldrich
Carbon Film Supported Copper Hexagonal Mesh Grid
size 200 mesh, box of 50
Synonym(s):
CARBON FILM ONLY ON 200 HEX COPPER GRIDS 50/BX
About This Item
UNSPSC Code:
41111761
material
carbon film
copper grid
description
film layer thickness 5 - 6 nm carbon
packaging
box of 50
size
200 mesh
General description
A 5-6 nm thin film of pure carbon supported hexagonal mesh grids. Copper grid, 200 mesh. Carbon film show better electrical and heat conduction in the electron beam.
replaced by
Choose from one of the most recent versions:
Certificates of Analysis (COA)