Key Documents
TEM-CF150NIUL
Sigma-Aldrich
Ultrathin Carbon Film Supported Nickel Square Mesh
size 150 mesh, box of 50
Synonym(s):
CARBON SQUARE MESH - NI 150 UL 50/BX
About This Item
UNSPSC Code:
41111761
packaging
box of 50
size
150 mesh
General description
A 3-4 nm ultrathin film of pure carbon supported standard squre mesh grids. Nickel grid, 150 mesh.
Choose from one of the most recent versions:
Certificates of Analysis (COA)