Key Documents
TEM-CF100HCUUL
Sigma-Aldrich
Ultrathin Carbon Film Supported Copper Hexagonal Mesh Grid
size 100 mesh, box of 50
Synonym(s):
CARBON HEXAGONAL MESH - CU 100 MESH UL 50/BX
About This Item
UNSPSC Code:
41111761
packaging
box of 50
size
100 mesh
General description
A 3-4 nm ultrathin film of pure carbon supported hexagonal mesh grids. Copper grid, 100 mesh. Carbon film show better electrical and heat conduction in the electron beam.
Choose from one of the most recent versions:
Certificates of Analysis (COA)