Key Documents
TEM-7596406
Sigma-Aldrich
OMNI Probe Molybdenum 4-Post Lift-Out Grid
pack of 25 (per vial)
Synonym(s):
OMNI GRIDS, 4-Post Lift-Out Grids - Molybdenum
About This Item
UNSPSC Code:
41111761
packaging
pack of 25 (per vial)
diam.
3 mm
General description
Custom Mo 4-post lift-out grids for in-situ lift-out. Include multiple indexed mounting locations, two with vertical bars attachment surfaces and two with “V” shaped alignment surfaces. Sides have lower profile for easier access to outermost posts.
replaced by
Choose from one of the most recent versions:
Certificates of Analysis (COA)