Key Documents
TEM-7435704
Sigma-Aldrich
Carbon Film Supported Copper Tomography Grid R14C2
pack of 50 ×
Synonym(s):
Carbon Film on Tomography Grid, 50pk
About This Item
UNSPSC Code:
41111761
packaging
pack of 50 ×
General description
This 1.5mm square, 300 mesh grid is made from copper, supported by a carbon film, and is for use with Fischione tomography specimen holders. The size allows for further tilt in TEMs with small pole piece holes. An identification mark and the square shape give easy reference when rotated past 90°.
Choose from one of the most recent versions:
Certificates of Analysis (COA)