Key Documents
TEM-70944
Sigma-Aldrich
Perfect Loop Set for Ultra Thin sections
Synonym(s):
PERFECT LOOP SET R10C9
About This Item
UNSPSC Code:
41111761
General description
Set of Handle & Loop: Perfect Loop for Ultra Thin Sections. Using this PERFECT LOOP, you can place your thin sections, cut on the ultramicrotome, easily on the grid mesh without creases.
Choose from one of the most recent versions:
Certificates of Analysis (COA)