About This Item
Recommended Products
reaction suitability
reagent type: ligand
functional group
phosphine
SMILES string
CCCCCCCCc1ccc2c-3c(OP(O)(=O)Oc4c(cc5cc(CCCCCCCC)ccc5c-34)C6CCCCC6)c(cc2c1)C7CCCCC7
InChI
1S/C84H113O4P/c1-3-5-7-9-11-19-33-59-47-49-71-69(51-59)57-77(79-73(63-39-25-15-26-40-63)53-67(61-35-21-13-22-36-61)54-74(79)64-41-27-16-28-42-64)83-81(71)82-72-50-48-60(34-20-12-10-8-6-4-2)52-70(72)58-78(84(82)88-89(85,86)87-83)80-75(65-43-29-17-30-44-65)55-68(62-37-23-14-24-38-62)56-76(80)66-45-31-18-32-46-66/h47-58,61-66H,3-46H2,1-2H3,(H,85,86)
InChI key
PQFDAAYIQUVLDR-UHFFFAOYSA-N
Other Notes
NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
TRIP and TiPSY Chiral Phosphoric Acid Catalysts
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service