Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

T26506

Sigma-Aldrich

Tetraphenylmethane

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H20
CAS Number:
Molecular Weight:
320.43
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
Pricing and availability is not currently available.

assay

97%

SMILES string

c1ccc(cc1)C(c2ccccc2)(c3ccccc3)c4ccccc4

InChI

1S/C25H20/c1-5-13-21(14-6-1)25(22-15-7-2-8-16-22,23-17-9-3-10-18-23)24-19-11-4-12-20-24/h1-20H

InChI key

PEQHIRFAKIASBK-UHFFFAOYSA-N

Application

Used to prepare a tetrapyridone tecton that forms a diamondoid network with large internal chambers.[1]

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Journal of the American Chemical Society, 113, 4696-4696 (1991)
Silke B Bodendiek et al.
Frontiers in pharmacology, 3, 106-106 (2012-06-12)
The paucity of specific pharmacological agents has been a major impediment for delineating the roles of gap junction (GJ) channels formed by connexin proteins in physiology and pathophysiology. Here, we used the selective optimization of side activities (SOSA) approach, which

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service