Key Documents
SYNS630012
Synthware low hold up high vacuum valve with protected o-rings
size 0-12 mm
Synonym(s):
Synthware S630012
Select a Size
1 EA
$123.00
$123.00
Check Cart for Availability
Notify Me
Get notified when this item is ready to ship via email.
Select a Size
Change View
1 EA
$123.00
About This Item
UNSPSC Code:
40161501
NACRES:
NB.51
$123.00
Check Cart for Availability
Notify Me
Get notified when this item is ready to ship via email.
packaging
pkg of 1 ea
manufacturer/tradename
Synthware S630012
General description
With two 9 mm O.D. sidearms bent to minimize hold up of liquids, Viton® O-ring on PTFE shaft are protected with precision PTFE wipers. The plug has a machined-in integral PTFE tip O-ring to make the primary seal. This design provides for an excellent vacuum tight seal without exposing any material other than PTFE to chemicals
Legal Information
Synthware is a trademark of Kemtech America Inc.
Viton is a registered trademark of E. I. du Pont de Nemours and Company
Choose from one of the most recent versions:
Certificates of Analysis (COA)