Key Documents
SYNM522404
Synthware vacuum manifold with high vacuum valves
port size 4, Hose Connection: left
About This Item
UNSPSC Code:
40161501
NACRES:
NB.51
packaging
pkg of 1 ea
manufacturer/tradename
Synthware M522404
port size
4
General description
With a serrated hose connection(10 mm O.D.) at left hand side. Main tube is 22 mm O.D. and the distance between valves is 100 mm center to center.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)