Key Documents
SYNF687000A
Synthware Sure-Grip separatory funnel
1000 mL, stopper size #27
About This Item
UNSPSC Code:
41121809
NACRES:
NB.51
packaging
pkg of 1 ea
manufacturer/tradename
Synthware F687000A
capacity
1000 mL
General description
Globe shaped separatory funnel with a long stopper neck and straight section between the funnel body and stopcock, which provides a secure grip while shaking and also aids in clamping. Available with PTFE stopcocks. Supplied complete with one listed size PTFE or glass stopper.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)