Key Documents
SYNF324100L
Synthware pear shaped flask
100 mL, joint: ST/NS 24/40
Synonym(s):
Synthware F324100L, pear shape glass flask
About This Item
UNSPSC Code:
41121804
NACRES:
NB.51
material
borosilicate glass flask
pear-shaped
single-neck
joint
joint: ST/NS24/40
packaging
pkg of 1 ea
manufacturer/tradename
Synthware F324100L
capacity
100 mL
100 mL
General description
Heavy wall design.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)