Key Documents
SYNC368030C
Synthware chromatography column with PTFE stopcock and coarse frit
L 305 mm
About This Item
UNSPSC Code:
41121800
NACRES:
NB.51
packaging
pkg of 1 ea
manufacturer/tradename
Synthware C368030C
L
305 mm
suitability
suitable for chromatography
General description
With reinforced beaded top, coarse porosity fritted disc to support column packing. All Synthware columns are heavy wall designed to increase safety.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)