Key Documents
SYNC184461C
Synthware chromatography column with PTFE stopcock and coarse fritted disc
top joint: ST/NS 24/40, I.D. × L 40.0 mm × 203 mm
About This Item
UNSPSC Code:
41121800
joint
joint: ST/NS top24/40
packaging
pkg of 1 ea
manufacturer/tradename
Synthware C184461C
I.D. × L
40.0 mm × 203 mm
General description
General purpose column has a PTFE stopcock and a top outer joint. With a coarse porosity fritted disc to support column packing. The section below disc is carefully minimized to prevent re-mixing of components already separated. All Synthware columns are heavy wall designed to increase safety.
This product includes coarse fritted disc.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)