Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SLWSSV304

Pyrex® Stopcocks, high vacuum

T-Key bore, bore 4 mm , key

Synonym(s):

Pyrex, SciLabware SSV304

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.43

manufacturer/tradename

SSV304
SciLabware

bore

4 mm , key

side arm thickness × O.D.

1.5 mm × 10 mm

Looking for similar products? Visit Product Comparison Guide

General description

  • Pyrex borosilicate glass construction
  • Each key and barrel are numbered to ensure they are always paired together
  • All side-arms are 100mm in length

Legal Information

PYREX is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service