SLW3730/04
Pyrex® Filter funnel with sintered glass disc
20 mL, bowl I.D. 65 mm, porosity grade 2
Synonym(s):
Pyrex, SciLabware 3730/04
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41104900
NACRES:
NB.43
Recommended Products
material
borosilicate glass
manufacturer/tradename
3730/04
SciLabware
bowl I.D.
65 mm
capacity
20 mL
20 mL
stem O.D. × L
8 mm × 75 mm
pore size
grade 2 porosity
Looking for similar products? Visit Product Comparison Guide
General description
- Sintered disc manufactured from Pyrex borosilicate glass
- Available in a choice of porosities
For the filtration of solutions:
- No requirement for filter paper
- Resistant to most corrosive reagents including ammonia and concentrated sulphuric acid
Legal Information
PYREX is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service