Key Documents
SLW3400/04
Pyrex® Crow receiver, class B
Synonym(s):
Pyrex, SciLabware 3400/04
About This Item
UNSPSC Code:
41121803
NACRES:
NB.43
Product SLW3400/04 is not currently sold in your country. Contact Technical Service
manufacturer/tradename
3400/02
SciLabware
General description
  • Complies with BS 658
  • For the collection of distillate in oil and tar tests
  • Graduations in white enamel
  • Manufactured from Pyrex borosilicate glass
Legal Information
PYREX is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)