Key Documents
SLW1522/12D
Pyrex® Stoppers, solid glass
joint: 60/46
Synonym(s):
Pyrex, SciLabware 1522/12D
About This Item
UNSPSC Code:
41122500
joint
cone joint 60/46
joint: 60/46
manufacturer/tradename
1522/12D
SciLabware
head diam.
94 mm
General description
  • Ideally suited for use with Pyrex bottles and aspirators
  • Robust, solid glass construction
  • Hexagonal head to provide a good grip
Legal Information
PYREX is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)