Key Documents
SLW1522/02D
Pyrex® Stoppers, solid glass
joint: 14/15
Synonym(s):
Pyrex, SciLabware 1522/02D
About This Item
UNSPSC Code:
41122500
joint
cone joint 14/15
joint: 14/15
manufacturer/tradename
1522/02D
SciLabware
head diam.
32 mm
General description
  • Ideally suited for use with Pyrex bottles and aspirators
  • Robust, solid glass construction
  • Hexagonal head to provide a good grip
Legal Information
PYREX is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)