Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SIK7407-10.4

Sigma-Aldrich

Hydria

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
$465.00

$465.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Request a Bulk Order

Select a Size

Change View
100 ML
$465.00

About This Item

UNSPSC Code:
12352103
NACRES:
NA.23

$465.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Request a Bulk Order

Application

Hydrophillic treatment for metals, metal oxides and alloys. Solution for nanocoating used to create hydrophilic surfaces.

Legal Information

Product of SiKEMIA

pictograms

FlameExclamation mark

signalword

Danger

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

target_organs

Central nervous system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

53.6 °F

flash_point_c

12 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service