Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SIK4405-30

Sigma-Aldrich

11-Pentafluorophenoxyundecyltriethoxysilane

≥95%

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
$156.00
1 G
$465.00

$156.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Request a Bulk Order
Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
250 MG
$156.00
1 G
$465.00

About This Item

Empirical Formula (Hill Notation):
C23H37F5O4Si
CAS Number:
Molecular Weight:
500.61
UNSPSC Code:
12352103
NACRES:
NA.23

$156.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Request a Bulk Order
Notify Me

Get notified when this item is ready to ship via email.

assay

≥95%

form

liquid (colorless)

shelf life

3 years under the recommended conditions

application

This coupling agent is an "hydrophobic ether" and gives very good results in the non-covalent immobilization (hydrophobic and pi stacking interactions) of proteins like antibodies.

Legal Information

Product of SiKEMIA

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service