Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

S6253

Sigma-Aldrich

Sodium phosphate glass

Type 65

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352300

mol wt

65 P±5 P (av. chain length)

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Reported to be of use in protein binding and sequestering and in studies on bone calcification.

Other Notes

Nan+2PnO3n+1; n = the average number of phosphorus (P) atoms in the chain.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service