RNI00183
(t-Bu)2 MeP Pd G2
AldrichCPR
Synonym(s):
(t-Bu)2 MeP Pd G2, Methanesulfonato (di-tert-butyl) methylphosphino (2′-amino-1,1′-biphenyl-2-yl) palladium(II)
About This Item
Recommended Products
form
solid
feature
generation 2
reaction suitability
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Coupling Reactions
reaction type: Cross Couplings
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst
functional group
phosphine
SMILES string
CP(C(C)(C)C)C(C)(C)C.NC1=C(C2=C([Pd]Cl)C=CC=C2)C=CC=C1
InChI
1S/C12H10N.C9H21P.ClH.Pd/c13-12-9-5-4-8-11(12)10-6-2-1-3-7-10;1-8(2,3)10(7)9(4,5)6;;/h1-6,8-9H,13H2;1-7H3;1H;/q;;;+1/p-1
InChI key
CPRPKFWSXAUWNS-UHFFFAOYSA-M
Other Notes
NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Buchwald precatalysts and ligands are bulky electron-rich dialkylbiaryl phospine-based catalysts and structurally related ligands that improve reactivity in Pd-catalyzed cross - coupling reactions
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service