Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

RNI00015

Sigma-Aldrich

2,6-Bis(diphenylmethyl)-4-methylaniline

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C33H29N
Molecular Weight:
439.59
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

SMILES string

Cc1cc(C(c2ccccc2)c3ccccc3)c(N)c(c1)C(c4ccccc4)c5ccccc5

InChI

1S/C33H29N/c1-24-22-29(31(25-14-6-2-7-15-25)26-16-8-3-9-17-26)33(34)30(23-24)32(27-18-10-4-11-19-27)28-20-12-5-13-21-28/h2-23,31-32H,34H2,1H3

InChI key

NCXCFZUKRMOFHW-UHFFFAOYSA-N

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service