Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

QBD10424

Sigma-Aldrich

m-dPEG®12-Propionaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H52O13
CAS Number:
Molecular Weight:
572.68
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

assay

>90%

form

solid or viscous liquid

reaction suitability

reagent type: chemical modification reagent
reagent type: cross-linking reagent
reactivity: carboxyl reactive

polymer architecture

shape: linear
functionality: monofunctional

shipped in

ambient

storage temp.

−20°C

SMILES string

COCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCC([H])=O

InChI

1S/C6H12O3/c1-8-5-6-9-4-2-3-7/h3H,2,4-6H2,1H3

InChI key

PVMNWBRYHQDVIN-UHFFFAOYSA-N

Features and Benefits

The m-dPEG®12-propionaldehyde consists of a methoxy-terminated, single molecular weight dPEG® molecule functionalized with a propionaldehyde group. The propionaldehyde moiety can be conjugated stably to amines, hydroxylamines, hydrazines, hydrazides, and other groups. The single molecular weight, hydrophilic, non-immunogenic m-dPEG® molecules improve water solubility, increase hydrodynamic volume, and decrease immunogenicity of target molecules.

Legal Information

Products Protected under U.S. Patents # 7,888,536 B2
dPEG is a registered trademark of Quanta BioDesign

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service