Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

QBD10204

Sigma-Aldrich

TFP-dPEG®12-biotinidase resistant biotin

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$345.00
1000 MG
$2,250.00

$345.00


Available to ship onApril 29, 2025Details


Request a Bulk Order

Select a Size

Change View
100 MG
$345.00
1000 MG
$2,250.00

About This Item

Empirical Formula (Hill Notation):
C47H76F4N4O17S
Molecular Weight:
1077.18
UNSPSC Code:
51171641
PubChem Substance ID:
NACRES:
NA.22

$345.00


Available to ship onApril 29, 2025Details


Request a Bulk Order

assay

>90%

form

solid or viscous liquid

reaction suitability

reagent type: cross-linking reagent
reaction type: Biotinylations
reactivity: amine reactive

reagent type: linker

polymer architecture

shape: linear
functionality: heterobifunctional

shipped in

ambient

storage temp.

−20°C

SMILES string

O=C1NC(C2N1)CSC2CCCCC(NC(C(NCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCC(OC3=C(C(F)=CC(F)=C3F)F)=O)=O)CC)=O

InChI key

NWCIJXJOOMRCER-UHFFFAOYSA-N

Features and Benefits

TFP-dPEG®12-biotinidase resistant biotin contains an amine reactive TFP ester and chemically modified biotin that is stable in the presence of biotinidase. Resistance to biotinidase cleavage of biotin is necessary in some biotechnology applications. The chemical modification to biotin imparts resistance to biotinidase cleavage without reducing the strength of biotin-avidin binding. The single molecular weight dPEG® spacer imparts water solubility, prevents aggregation and precipitation of labeled peptides and proteins, and reduces non-specific binding. Two different single molecular weight dPEG® spacers are available for precise spatial control between the modified biotin and the surface of the molecule to which it is conjugated.

Legal Information

Products Protected under U.S. Patents # 7,888,536 B2
dPEG is a registered trademark of Quanta BioDesign

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service