P4402
3-Pentadecylphenol
technical grade, 90%
Synonym(s):
2-Deoxy-urushiol I
About This Item
Recommended Products
grade
technical grade
assay
90%
bp
190-195 °C/1 mmHg (lit.)
mp
50-53 °C (lit.)
SMILES string
CCCCCCCCCCCCCCCc1cccc(O)c1
InChI
1S/C21H36O/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-16-20-17-15-18-21(22)19-20/h15,17-19,22H,2-14,16H2,1H3
InChI key
PTFIPECGHSYQNR-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
signalword
Warning
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
235.4 °F - closed cup
flash_point_c
113 °C - closed cup
ppe
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service