Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

OCNQAK560

Sigma-Aldrich

SMCC Conjugation Kit for Amine Cd-based Core/Shell Quantum Dots

fluorescence λem 560 nm, for 4 conjugation

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
43211915

fluorescence

λem 560 nm

Looking for similar products? Visit Product Comparison Guide

General description

Components:
  • QSA
  • Sulfo-SMCC
  • Traut′s Reagent
  • Coupling Buffer
  • Storage Buffer
  • Quenching Buffer
  • Tubes 4 x 1.7 mL

Legal Information

Product of Ocean Nanotech, LLC.

pictograms

Skull and crossbonesEnvironment

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Chronic 2

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Cadmium-free quantum dots for bioimaging offer design and synthesis strategies, enhancing bioimaging and sensing applications.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service