O9220
Oxamic acid ammonium salt
Synonym(s):
Ammonium oxamate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
H2NC(O)CO2H · NH3
Molecular Weight:
106.08
MDL number:
UNSPSC Code:
41116105
PubChem Substance ID:
SMILES string
[NH4+].NC(=O)C([O-])=O
InChI
1S/C2H3NO3.H3N/c3-1(4)2(5)6;/h(H2,3,4)(H,5,6);1H3
InChI key
NGARWWRAFUCMAE-UHFFFAOYSA-N
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service