Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

O5007

Sigma-Aldrich

16-Oxokahweol

≥98% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H22O2
CAS Number:
Molecular Weight:
282.38
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

assay

≥98% (HPLC)

form

solid

storage temp.

−20°C

SMILES string

CC12C=Cc3occc3C1CCC45CC(CCC24)C(=O)C5

InChI

1S/C19H22O2/c1-18-7-5-16-13(6-9-21-16)14(18)4-8-19-10-12(15(20)11-19)2-3-17(18)19/h5-7,9,12,14,17H,2-4,8,10-11H2,1H3/t12-,14+,17-,18+,19+/m0/s1

InChI key

ZUHATORLUREVSD-OXQYBPPISA-N

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L K Lam et al.
Journal of medicinal chemistry, 30(8), 1399-1403 (1987-08-01)
The coffee constituents cafestol and kahweol are inducers of the activity of the detoxifying enzyme, glutathione S-transferase in laboratory animals. The two active functional groups, furan and glycol, on opposite ends of the diterpene nucleus of these two compounds have

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service