Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

O202

Octachlorocyclopentene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5Cl8
Molecular Weight:
343.68
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
Pricing and availability is not currently available.

SMILES string

ClC1=C(Cl)C(Cl)(Cl)C(Cl)(Cl)C1(Cl)Cl

InChI

1S/C5Cl8/c6-1-2(7)4(10,11)5(12,13)3(1,8)9

InChI key

DMZRCHJVWAKCAX-UHFFFAOYSA-N


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service