Key Documents
NOR5X425INSB
Optimizer insert for spinner turbines
NMR tube size 4.25 mm , into 5 mm Bruker
About This Item
UNSPSC Code:
41121705
NACRES:
SB.32
material
(acetal homopolymer resin)
manufacturer/tradename
Norell
NMR tube size
4.25 mm , into 5 mm Bruker
General description
Precision adapter holds 3 mm, 4 mm, and 4.25 mm NMR tube in a 5 mm spinner turbine. Does not include spinner turbine.
Choose from one of the most recent versions:
Certificates of Analysis (COA)