Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

NCXSEPD5

Sigma-Aldrich

Silver Nanospheres

5 nm, Econix, Dried, PVP 40 kDa (Polymer), powder

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12162002
NACRES:
NA.23

form

nanospheres
powder

packaging

pkg of SKU size by silver mass

particle size

5 nm

Looking for similar products? Visit Product Comparison Guide

Application

Owing to their plasmonic properties nanoparticles of gold and silver find broad range of applications extending from diagnostics to molecular detection, photothermal and microwave-based therapies, and nanotoxicology. Colloidal silver nanoparticles also find applications in antimicrobial coatings, electronics and photovoltaics, and surface enhanced Raman spectroscopy (SERS) whereas the colloidal gold nanoparticles find applications in lateral flow assays (immunochromatographic assays) and as TEM labels and size standards.

Our Econix nanospheres contain PVP (40 kD polyvinylpyrrolidone) as excipient. The PVP enhances stability and dispersibility of Econix silver nanospheres. These dried powders are redispersible in a variety of polar solvents such as water, alcohols, DMF, and DMSO. However, they show medium dispersibility in chloroform, and low dispersibility in dichloromethane. Econix materials are unagglomerated, low cost silver nanospheres and are ideal for applications requiring large quantities.

Storage and Stability

DO NOT FREEZE. Store at 2-25C in a dry place. Nanomaterials can be sensitive to high and low pH conditions and high salt concentrations. Use caution when changing environmental conditions as particles may aggregate.

Legal Information

Product of nanoComposix, Inc.

pictograms

Environment

signalword

Warning

hcodes

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Research on noble bimetallic nanoparticles for glucose biosensor detection.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service