NCXSEPD110
Silver Nanospheres
110 nm, Econix, Dried, PVP 40 kDa (Polymer), powder
About This Item
form
nanospheres
powder
packaging
pkg of SKU size by silver mass
particle size
110 nm
Looking for similar products? Visit Product Comparison Guide
Application
Our Econix nanospheres contain PVP (40 kD polyvinylpyrrolidone) as excipient. The PVP enhances stability and dispersibility of Econix silver nanospheres. These dried powders are redispersible in a variety of polar solvents such as water, alcohols, DMF, and DMSO. However, they show medium dispersibility in chloroform, and low dispersibility in dichloromethane. Econix materials are unagglomerated, low cost silver nanospheres and are ideal for applications requiring large quantities.
Storage and Stability
Legal Information
signalword
Warning
hcodes
pcodes
Hazard Classifications
Aquatic Acute 1 - Aquatic Chronic 1
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Research on noble bimetallic nanoparticles for glucose biosensor detection.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service