NCXAUKR80
Gold Nanospheres
Bare, 80 nm, for Passive Adsorption, Bioready, Carbonate, OD 5, in Aqueous Potassium Carbonate
About This Item
Recommended Products
form
nanospheres
OD
5
particle size
80 nm
Looking for similar products? Visit Product Comparison Guide
Application
The BioReady gold nanospheres are well suited for passive conjugation of antibodies, proteins, peptides, and other biomolecules.
The BioReady citrate gold nanoparticles have a “naked” particle surface which contains a weakly bound citrate molecule as stabilizer. The citrate is readily displaced in presence of proteins and allows passive adsorption (also referred to as physisorption). The mechanism of adsorption is based on Van der Waals interactions between the proteins (e.g. antibodies) and the surface of the particles. The resulting forces between the antibody and the nanoparticle are influenced by the coupling environment. Our BioReady citrate gold is provided at an optical density (OD) of 20 at pH ~8.0–8.5. A pH titration should be performed to optimize the pH of conjugation.
The BioReady carbonate gold nanoparticles have a “naked” particle surface that contains a weakly bound carbonate molecule as particle stabilizer and, similar to citrate-stabilized gold, the carbonate is readily displaced in the presence of proteins and allows easy passive adsorption. Carbonate is a smaller and less complex molecule and has a lower affinity to the gold nanoparticle surface compared to citrate. This enhances the displaceability of the carbonate molecules allows better conjugation. Our BioReady carbonate gold is provided at an optical density (OD) of 20 at pH ~7.4–8.6. A pH titration should be performed to optimize the pH of conjugation.
Storage and Stability
Legal Information
Storage Class
12 - Non Combustible Liquids
wgk_germany
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service