NCXAGAH70
Silver Nanospheres
Silica-Shelled, 70 nm, NanoXact, 1 mg/mL in ethanol, Aminated / Cationic Silica
About This Item
form
nanospheres
concentration
1 mg/mL in ethanol
particle size
70 nm
Looking for similar products? Visit Product Comparison Guide
Application
Our silica shelled silver nanospheres possess robust surface and exhibit high stability in variety of solvents. The silica shell offers a versatile surface for attaching functional groups or creating hydrophobic & hydrophilic surfaces. The presence of silica shell on the surface of silver nanospheres also allows their easier deposition onto substrates and incorporation into different solvents, resins, and curable materials while protecting them from aggregation and preserving their optical properties.
Storage and Stability
Legal Information
signalword
Danger
hcodes
Hazard Classifications
Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Flam. Liq. 2
Storage Class
3 - Flammable liquids
wgk_germany
WGK 3
flash_point_f
55.4 °F
flash_point_c
13 °C
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Research on noble bimetallic nanoparticles for glucose biosensor detection.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service