Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

N6005

Sigma-Aldrich

2-Naphthyl phosphate disodium salt

2-Naphthyl phosphate disodium salt
1 of 1 reviewers received a sample product or took part in a promotion

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C10H7OP(O)(ONa)2
CAS Number:
Molecular Weight:
268.11
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

97%

form

powder

storage temp.

−20°C

SMILES string

O.[Na+].[Na+].[O-]P([O-])(=O)Oc1ccc2ccccc2c1

InChI

1S/C10H9O4P.2Na.H2O/c11-15(12,13)14-10-6-5-8-3-1-2-4-9(8)7-10;;;/h1-7H,(H2,11,12,13);;;1H2/q;2*+1;/p-2

InChI key

VZEQFWJGQGFKLL-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

2-Naphthyl phosphate disodium salt has been used to study its binding characteristics onto alumina.[1]

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Adsorption characteristics of charged and nonionic small molecules to colloidal alumina
Pansare VJ, et al.
Journal of Colloid and Interface Science, 512, 29-38 (2018)
A Buist et al.
Biochemistry, 38(3), 914-922 (1999-01-20)
Most transmembrane, receptor-like protein-tyrosine phosphatases (RPTPs) contain two cytoplasmic catalytic protein-tyrosine phosphatase (PTP) domains, of which the membrane-proximal domain, D1, contains the majority of the activity, while the membrane-distal domain, D2, exhibits little or no activity. We have investigated the
José Mauro Granjeiro et al.
Journal of enzyme inhibition and medicinal chemistry, 17(5), 345-350 (2003-04-10)
Uric acid inhibited 50% of the activity of bovine kidney low molecular mass phosphotyrosine protein phosphatase at concentrations of 1.0, 0.4, 1.3, and 0.2 mM, respectively for p-nitrophenyl phosphate (p-NPP), flavine mononucleotide, beta-naphthyl phosphate and tyrosine phosphate (Tyr-P) as substrates.
J M Granjeiro et al.
Biochemistry and molecular biology international, 41(6), 1201-1208 (1997-05-01)
A low molecular weight bovine kidney acid phosphatase, electrophoretically homogeneous and with a relative molecular mass of 17.8 kDa, was used in this work. Among the various substrates tested, FMN was found to be the most effective, at pH 7.0.
C Hipólito-Reis et al.
Scandinavian journal of clinical and laboratory investigation, 59(8), 593-606 (2000-02-26)
The importance of separation and identification of serum alkaline phosphatase (ALP; E.C. 3.1.3.1) fractions/isoenzymes has been frequently reported. Each serum ALP fraction/isoenzyme quantitation has both practical and theoretical importance. In the present work, serum was collected from Wistar rats and

Questions

Reviews

1 of 1 reviewers received a sample product or took part in a promotion

Active Filters

  1. 1 Ratings-Only Review

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service