Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

N16401

Sigma-Aldrich

trans-2-Nitrocinnamic acid

98%

Synonym(s):

trans-3-(2-Nitrophenyl)acrylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
O2NC6H4CH=CHCO2H
CAS Number:
Molecular Weight:
193.16
Beilstein/REAXYS Number:
2211209
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

98%

form

crystals

mp

243-245 °C (lit.)

SMILES string

OC(=O)\C=C\c1ccccc1[N+]([O-])=O

InChI

1S/C9H7NO4/c11-9(12)6-5-7-3-1-2-4-8(7)10(13)14/h1-6H,(H,11,12)/b6-5+

InChI key

BBQDLDVSEDAYAA-AATRIKPKSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service