Key Documents
MT59902917
Mettler Toledo RedCap combined pH Electrode for flow-through cell
L 40 mm × 12 mm
About This Item
UNSPSC Code:
41115604
NACRES:
NB.79
Pricing and availability is not currently available.
material
glass (U)
glass membrane (polymer)
description
S7 connection
Without cable
manufacturer/tradename
Mettler-Toledo 59902917
L × W
40 mm × 12 mm
L
40 mm × 12 mm
General description
Mettler Toledo RedCap combined pH Electrode U402-611-S7/40 is generally used in conjunction with the flow-through cell 611 (59904354). This enables high precision measurements of various samples. This electrode features a double ceramic diaphragm and a low membrane resistance (< 250 M?).
  • Ideal for measurements of small sample volumes and samples with low ionic strength
  • Ground glass fitting for flow cell 611 (59904354)
  • Double ceramic junction with pore size 0.9 μm
  • SteadyForce reference system
Legal Information
SteadyForce is a trademark of Mettler-Toledo, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)