Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MT52000379

Mettler-Toledo InLab® pH electrode 413/IP67

for use with emulsions, suspensions, effluent water and solutions containing proteins and sulphides

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115604
NACRES:
NB.79
Pricing and availability is not currently available.

description

BNC/RCA (Cinch), IP67 connector

manufacturer/tradename

Mettler-Toledo 52000379

General description

Probe is fully protected against ingress of water and dust (IP67 rated).

Legal Information

InLab is a registered trademark of Mettler-Toledo, Inc.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service