Key Documents
MT51344612
Mettler Toledo InLab® 605-ISM dissolved oxygen sensor
cable L 5 m
About This Item
UNSPSC Code:
39121436
NACRES:
NB.79
Pricing and availability is not currently available.
description
0 - 200%, 0 - 20 mg/L
BNC/RCA (Cinch), ISM connector
manufacturer/tradename
Mettler-Toledo 51344612
cable L
5 m
General description
The InLab® 605-ISM-5m DO sensor incorporates the newest technology in polarographic dissolved oxygen determination. With theInLab® 605-ISM-5m dissolved oxygen sensor you will find the high accuracy and reproducibility that you have come to trust with METTLER TOLEDO products.
The InLab® 605-ISM-5m sensor can be used in combination with a SevenExcellence multichannel benchtop meter or the SevenGo 68 dual channel portable meters.
Features and Benefits
There are certain applications in which such sensors need to be rugged enough to withstand harsh conditions in factories, laboratories or out in the field. For this reason the membrane is protected by a steel mesh and the shaft is made out of extremely resistant glass fibre reinforced PPS (polyphenylene sulfide).
To go with the InLab® 605-ISM-5m METTLER TOLEDO offers the best of the best accessories: membrane kit, electrolyte, sealing kit and electrode weight.
Legal Information
InLab is a registered trademark of Mettler-Toledo, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)