Key Documents
MT51343202
Mettler Toledo InLab® Redox-L ORP electrode
Extra Long Redox Electrode with Platinum Ring
About This Item
UNSPSC Code:
39121436
NACRES:
NB.79
Pricing and availability is not currently available.
material
glass shaft
description
S7 connection
Without cable
manufacturer/tradename
Mettler-Toledo 51343202
temperature measuring range
0-100 °C
L × W
170 mm × 6 mm
General description
Mettler Toledo InLab® Redox –L is a combination electrode for measuring the oxidation/reduction power of a solution. Consists of a platinum ring indicator electrode and a silver/silver chloride reference electrode.
The measuring signal is produced on the surface of the precious metal by an exchange of electrons with the oxidation-reduction system of the measured medium.
Combination design avoids use of separate reference and indicating electrodes.
The innovative Safelock closing ring gives easy access for re-filling reference electrolyte and complete sealing when closed. Different cables can be connected to the S7 screw head allowing use with a wide range of meters.
• 170 mm shaft length
• Combination design
• Safelock closing ring
• Detachable cable
Legal Information
InLab is a registered trademark of Mettler-Toledo, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)