Key Documents
MT51343195
Mettler Toledo InLab® Mono pH half-cell electrode
pH half-cell, glass, HA membrane
About This Item
UNSPSC Code:
41115604
NACRES:
NB.79
Pricing and availability is not currently available.
material
glass shaft
glass, HA membrane
description
S7 connector
Without cable
manufacturer/tradename
Mettler-Toledo 51343195
L × W
120 mm × 12 mm
pH range
0-14
General description
InLab® Mono series are half-cell glass-body, pH electrodes with S7 screw head connector.
Recommended for applications where the life time of the pH electrode is significantly shorter than the reference electrode.
The use of a separate pH electrode can save money for those applications where lifetime of the reference electrode is much longer than the pH electrode.
Glass body gives protection against corrosive and organic samples. Different cables can be connected to the S7 screw head allowing use with a wide range of meters.
  • Extended reference lifetime
  • Detachable cable
  • Glass body
Mono
Legal Information
InLab is a registered trademark of Mettler-Toledo, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)