MT51343062
Mettler Toledo InLab® Reach pH electrode
MultiPin™ connector, L × W 400 mm × 12 mm, Temperature probe Pt1000
About This Item
Recommended Products
material
glass shaft
glass, HA membrane
description
without cable
manufacturer/tradename
Mettler-Toledo 51343062
temperature measuring range
0-100 °C
L × W
400 mm × 12 mm
pH
1-14
Looking for similar products? Visit Product Comparison Guide
General description
Designed for accurate measurement in a wide range of sample types.
The electrolyte in conventional silver/silver chloride reference systems must be saturated with silver chloride. With critical media such as those containing sulphides or proteins, this will result in diaphragm contamination with silver sulphide. This can not happen with InLab® electrodes which have an ARGENTHAL™ lead off and silver ion trap that ensure the electrolyte remains completely free from silver ions.
The ARGENTHAL™ reference system also improves service life and measurement accuracy at differing sample temperatures.
The glass shaft provides resistance against organic and corrosive samples.
- ARGENTHAL™ reference with silver ion trap
- Glass shaft
- Detachable cable
- Extended length shaft
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service