MT51343031
Mettler-Toledo InLab® Versatile & Versatile Pro pH electrode
Temperature probe NTC 30 kOhm
About This Item
Recommended Products
material
glass, U membrane
polysulfone shaft
description
BNC/RCA (Cinch) connection
Reference electrolyte 3 mol/L KCl
Reference system ARGENTHAL™, with Ag+ -trap
manufacturer/tradename
Mettler-Toledo 51343031
temperature measuring range
0-100 °C
L × diam.
120 mm × 12 mm
cable L
1.2 m
pH range
0-14
Looking for similar products? Visit Product Comparison Guide
General description
Features and Benefits
- Temperature range 0-100 °C
- Replaceable ceramic junction
- Robust polysulfone shaft
- ARGENTHAL™ and silver ion trap
- BNC/RCA (Cinch) cable
Legal Information
related product
Storage Class
12 - Non Combustible Liquids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service